Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MVK Antibody (OTI 1D7), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272837UV
Description
MVK Monoclonal specifically detects MVK in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MVK | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MVK | |
Full length recombinant protein of human MVK (NP_000422) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI 1D7 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
EC 2.7.1.36, LH receptor mRNA-binding protein, LRBP, mevalonate kinase, mevalonate kinase (mevalonic aciduria), mevalonate kinase 1, MKFLJ96772, MVLK | |
Mouse | |
Affinity Purified | |
metabolism | |
4598.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction