Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MVK Monoclonal Antibody (OTI 1D7)
GREENER_CHOICE

Catalog No. PIMA525553
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525553 Supplier Invitrogen™ Supplier No. MA525553
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MVK Monoclonal Antibody for Western Blot, ICC/IF

Mevalonate kinase (MVK) is an early enzyme in isoprenoid and sterol synthesis. Mevalonate kinase catalyzes the ATP-dependent phosphorylation of mevalonic acid to form mevalonate 5-phosphate. Deficiency in MVK activity contributes to mevalonic aciduria and hyperimmunoglobulinemia D/periodic fever syndrome (HIDS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MVK
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI 1D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Mvk
Gene Accession No. Q03426
Gene Alias 2310010A05Rik; AI256848; AI414037; KIME; LH receptor mRNA-binding protein; LHR mRNA binding protein; Lrbp; Mevalonate kinase; mevalonate kinase 1; MK; Mvk; MVLK; POROK3
Gene Symbols Mvk
Host Species Mouse
Immunogen Full length human recombinant protein of MVK produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4598
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.