Learn More
MVK Mouse anti-Human, Clone: OTI 1D7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502934

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes the peroxisomal enzyme mevalonate kinase. Mevalonate is a key intermediate, and mevalonate kinase a key early enzyme, in isoprenoid and sterol synthesis. Mevalonate kinase deficiency caused by mutation of this gene results in mevalonic aciduria, a disease characterized psychomotor retardation, failure to thrive, hepatosplenomegaly, anemia and recurrent febrile crises. Defects in this gene also cause hyperimmunoglobulinaemia D and periodic fever syndrome, a disorder characterized by recurrent episodes of fever associated with lymphadenopathy, arthralgia, gastrointestinal dismay and skin rash. Two transcript variants that encode the same protein have been found for this gene.Specifications
MVK | |
Monoclonal | |
Unconjugated | |
MVK | |
LRBP, MK, MVLK, POROK3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4598 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI 1D7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q03426 | |
MVK | |
Full length human recombit protein of human MVK produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.