Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MVP Antibody (1014), PE , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311555PE
Description
MVP Monoclonal antibody specifically detects MVP in Human samples. It is validated for Flow CytometrySpecifications
MVP | |
Monoclonal | |
PE | |
LRPVAULT1, Lung resistance-related protein, major vault protein | |
proteins precipitated from human breast cancer MCF-7 cells | |
0.1 mL | |
Cancer, Signal Transduction | |
9961 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
1014 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction