Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MVP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131295
Description
MVP Polyclonal specifically detects MVP in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
MVP | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
MVP | |
Recombinant protein encompassing a sequence within the C-terminus region of human MVP/LRP. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Knockout Validated | |
LRPVAULT1, Lung resistance-related protein, major vault protein | |
Rabbit | |
99 kDa | |
100 μL | |
Cancer | |
9961 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction