Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MVP Antibody (SPM280), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311472H
Description
MVP Monoclonal antibody specifically detects MVP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MVP | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
SPM280 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
LRPVAULT1, Lung resistance-related protein, major vault protein | |
proteins precipitated from human breast cancer MCF-7 cells | |
0.1 mL | |
Cancer, Signal Transduction | |
9961 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction