Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MVP Antibody (VP2897R) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308801
Description
MVP Monoclonal specifically detects MVP in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
MVP | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry-Paraffin : 1-2 ug/mL | |
LRPVAULT1, Lung resistance-related protein, major vault protein | |
Recombinant full-length human MVP protein | |
100 μg | |
Cancer, Signal Transduction | |
9961 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry (Paraffin) | |
VP2897R | |
Unconjugated | |
10mM PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction