Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MVP Monoclonal Antibody (1032), Biotin, Invitrogen™

Catalog No. PIMA513868
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513868 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513868 Supplier Thermo Scientific Supplier No. MA513868
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-13868 targets Major Vault Protein in FACS, GS, IHC (P), IP, and WB applications and shows reactivity with Human and Rat samples. The MA5-13868 immunogen is proteins precipitated with AER317 from the phosphocellulose column flow through of extract of human breast cancer MCF-7 cells.

This gene encodes the major vault protein which is a lung resistance-related protein. Vaults are multi-subunit structures that may be involved in nucleo-cytoplasmic transport. This protein mediates drug resistance, perhaps via a transport process. It is widely distributed in normal tissues, and overexpressed in multidrug-resistant cancer cells. The protein overexpression is a potentially useful marker of clinical drug resistance. This gene produces two transcripts by using two alternative exon 2 sequences; however, the open reading frames are the same in both transcripts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MVP
Applications Flow Cytometry, Gel Shift, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1032
Concentration 0.2 mg/mL
Conjugate Biotin
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene MVP
Gene Accession No. Q14764, Q62667
Gene Alias 2310009M24Rik; LRP; Lung resistance-related protein; major vault protein; MVP; MVP1; Testicular Secretory Protein Li 30; VAULT1; vRNP
Gene Symbols MVP
Host Species Mouse
Immunogen Proteins precipitated with AER317 from the phosphocellulose column flow through of extract of human breast cancer MCF-7 cells
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64681, 9961
Target Species Human, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.