Learn More
Invitrogen™ MVP Monoclonal Antibody (MVP-37)

Description
MVP-37 reacts with an internal epitope of MVP/LRP (p110), which is strongly overexpressed in various human non-P-glycoprotein MDR tumor cell lines, accordingly to an increase in the number of vault particles.
Specifications
Specifications
| Antigen | MVP |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | MVP-37 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with 0.7% BSA and 0.1% sodium azide |
| Gene | MVP |
| Gene Accession No. | Q14764 |
| Gene Alias | 2310009M24Rik; LRP; Lung resistance-related protein; major vault protein; MVP; MVP1; Testicular Secretory Protein Li 30; VAULT1; vRNP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.