Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MVP Mouse anti-Human, Clone: 1032, Abnova™

Catalog No. 89034507 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-507 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-507 Supplier Abnova Corporation Supplier No. MAB14424
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human MVP.

  • High specificity
  • High reactivity

Specifications

Antigen MVP
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone 1032
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human MVP.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA and 0.05% azide).
Gene Accession No. Q14764
Gene Alias LRP/VAULT1
Gene Symbols MVP
Host Species Mouse
Immunogen Proteins precipitated from human breast cancer MCF-7 cell line.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9961
Test Specificity Recognizes a protein of 104kDa-110kDa which characterized as MVP.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.