Learn More
MVP Mouse anti-Human, Clone: SPM280, Abnova™
Mouse monoclonal antibody raised against native human MVP.
Supplier: Abnova Corporation MAB13191
Description
This gene encodes the major vault protein which is a lung resistance-related protein. Vaults are multi-subunit structures that may be involved in nucleo-cytoplasmic transport. This protein mediates drug resistance, perhaps via a transport process. It is widely distributed in normal tissues, and overexpressed in multidrug-resistant cancer cells. The protein overexpression is a potentially useful marker of clinical drug resistance. This gene produces two transcripts by using two alternative exon 2 sequences; however, the open reading frames are the same in both transcripts. [provided by RefSeq]
Specifications
MVP | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
LRP/VAULT1 | |
Mouse | |
Protein A/G purification | |
RUO | |
9961 | |
Store at 4°C. | |
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
SPM280 | |
Mouse monoclonal antibody raised against native human MVP. | |
In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
MVP | |
Native purified MVP from human breast cancer MCF-7 cells. | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.