Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583132 Supplier Invitrogen™ Supplier No. PA583132
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PYRRRSQFSS RKYLKKEMNS FQQQPPPFGT VPPQMMFPPN WQGAEKDAAF LAKDFNFLTL NNQPPPGNRS QPRAMG Highest antigen sequence indentity to the following orthologs: Mouse - 28%, Rat - 29%.

Members of the Dynamin family include GTPase, microtubule-associated proteins that are involved in cellular trafficking, including microtubule bundling and endocytosis. Mx1, also known as MxA, an interferon (IFN)-induced protein, acquires a high degree of resistance to influenza A virus and the rhabdo-virus vesicular stomatitis virus (VSV), which suggests that Mx1 plays an active role against influenza virus and the rhabdovirus VSV. Mx1 is a cytoplasmic protein that is 63% identical to the Mx2 protein, which lacks antiviral activity. Mx2 is also known as MxB and is localized at the cytoplasmic face of nuclear pores. Mx2 expression is not interferon-dependent and this protein is thought to regulate the efficiency and/or kinetics of nuclear import, a function which may have been usurped by its antiviral relatives.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MX2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MX2
Gene Accession No. P20592
Gene Alias AI528743; interferon-induced GTP-binding protein Mx2; interferon-induced GTP-binding protein Mx3; interferon-regulated resistance GTP-binding protein MXB; MX dynamin like GTPase 2; MX dynamin-like GTPase 2; Mx2; Mx-2; Mx3; MXB; myxovirus (influenza virus) resistance 2; myxovirus (influenza virus) resistance 3; myxovirus resistance protein 2; myxovirus resistance protein 3; p78-related protein; protein MxB; second interferon-induced protein p78
Gene Symbols MX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MX2. Recombinant protein control fragment (Product #RP-95904).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4600
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.