Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MxA/Mx1 Antibody (OTI2G12), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP147859
Description
MxA/Mx1 Monoclonal specifically detects MxA/Mx1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MxA/Mx1 | |
Monoclonal | |
0.3 mg/mL | |
Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
MX1 | |
Full length human recombinant protein of human MX1 (NP_002453) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific for Homo sapiens myxovirus (influenza virus) resistance 1, interferon-inducible protein p78 (mouse) (MX1), transcript variant 2. | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2G12 | |
Unconjugated | |
human interferon-regulated resistance GTP-binding protein MXA10IFI-78Khomolog of murine (interferon-inducibleprotein p78), myxovirus (influenza virus) resistance 1, interferon-inducible protein p78(mouse) | |
Mouse | |
75.5 kDa | |
0.1 mL | |
Apoptosis | |
4599 | |
Human, Mouse, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction