Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MxA/Mx1 Antibody (OTI2G12), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272838G
Description
MxA/Mx1 Monoclonal specifically detects MxA/Mx1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MxA/Mx1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MX1 | |
Full length recombinant protein of human MxA/Mx1 (NP_002453) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2G12 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
human interferon-regulated resistance GTP-binding protein MXA10IFI-78Khomolog of murine (interferon-inducibleprotein p78), myxovirus (influenza virus) resistance 1, interferon-inducible protein p78(mouse) | |
Mouse | |
Affinity Purified | |
Apoptosis | |
4599.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction