Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MXD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517613
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other Mad family members at physiological levels.
MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.
Specifications
MXD1 | |
Polyclonal | |
Unconjugated | |
MXD1 | |
antagonizer of myc transcriptional activity; AW122478; BHLHC58; Mad; Mad1; Mad-1; max dimerization protein 1; Max dimerizer 1; MAX-binding protein; MGC104659; mxd1; OTTHUMP00000202211; Protein MAD; si:dkey-202n14.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17119, 362391, 4084 | |
-20°C | |
Liquid |
Western Blot | |
29 μg/mL | |
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
P50538, Q05195 | |
MXD1 | |
Synthetic peptide corresponding to the carboxy terminus region of Mad-1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction