Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MXD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541433
Description
Peptide sequence: MAAAVRMNIQ MLLEAADYLE RREREAEHGY ASMLPYNNKD RDALKRRNKS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 92%; Zebrafish: 100%.
MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.
Specifications
MXD1 | |
Polyclonal | |
Unconjugated | |
MXD1 | |
antagonizer of myc transcriptional activity; AW122478; BHLHC58; Mad; Mad1; Mad-1; max dimerization protein 1; Max dimerizer 1; MAX-binding protein; MGC104659; mxd1; OTTHUMP00000202211; Protein MAD; si:dkey-202n14.4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
17119, 4084 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Surface Plasmon Resonance | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P50538, Q05195 | |
MXD1 | |
Synthetic peptide directed towards the N-terminal of human MXD1 (aa 1-50). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction