Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MXD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551609
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551609 Supplier Invitrogen™ Supplier No. PA551609
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LCLEKLKGLV PLGPESSRHT TLSLLTKAKL HIKKLEDCDR KAVHQIDQLQ REQRHLKRQL EKLGIERIRM DSIGSTVSSE RSDSDREEID VDVESTDYLT GDLDWSSSSV SDSDERGSMQ SLGSDEGYSS TSI Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MXD1
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MXD1
Gene Accession No. Q05195
Gene Alias antagonizer of myc transcriptional activity; AW122478; BHLHC58; Mad; Mad1; Mad-1; max dimerization protein 1; Max dimerizer 1; MAX-binding protein; MGC104659; mxd1; OTTHUMP00000202211; Protein MAD; si:dkey-202n14.4
Gene Symbols MXD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MXD1. Recombinant protein control fragment (Product #RP-100691).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4084
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.