Learn More
MXD4 Mouse anti-Human, Clone: OTI5E6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808855

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the MAD gene family. The MAD genes encode basic helix-loop-helix-leucine zipper proteins that heterodimerize with MAX protein, forming a transcriptional repression complex. The MAD proteins compete for MAX binding with MYC, which heterodimerizes with MAX forming a transcriptional activation complex. Studies in rodents suggest that the MAD genes are tumor suppressors and contribute to the regulation of cell growth in differentiating tissues.Specifications
MXD4 | |
Monoclonal | |
Unconjugated | |
MXD4 | |
MAD4, MST149, MSTP149, bHLHc12 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10608 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5E6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q14582 | |
MXD4 | |
Full length human recombit protein of human MXD4 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.