Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mxi1 Antibody (PCRP-MXI1-1A3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308353F
Description
Mxi1 Monoclonal antibody specifically detects Mxi1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
Mxi1 | |
Monoclonal | |
FITC | |
BHLHC11, Class C basic helix-loop-helix protein 11, MAX dimerization protein 2, MAX interacting protein 1, MAX interactor 1bHLHc11MAD2, max-interacting protein 1, Max-related transcription factor, MGC43220, MXD2, MXI | |
Recombinant full-length human Mxi1 protein (Uniprot: P50539) | |
100 μg | |
Cell Cycle and Replication | |
4601 | |
Store at 4°C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-MXI1-1A3 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction