Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mxi1 Mouse anti-Human, Clone: PCRP-MXI1-1A3, Novus Biologicals™

Mouse Monoclonal Antibody
$235.84
Specifications
| Antigen | Mxi1 |
|---|---|
| Clone | PCRP-MXI1-1A3 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array |
Description
Mxi1 Monoclonal specifically detects Mxi1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| Mxi1 | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| 10 mM PBS with 0.05% BSA | |
| 4601 | |
| IgG2b | |
| Protein A or G purified |
| PCRP-MXI1-1A3 | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BHLHC11, Class C basic helix-loop-helix protein 11, MAX dimerization protein 2, MAX interacting protein 1, MAX interactor 1bHLHc11MAD2, max-interacting protein 1, Max-related transcription factor, MGC43220, MXD2, MXI | |
| Recombinant full-length human Mxi1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title