Learn More
Invitrogen™ MXI1 Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is >95% (by SDS-PAGE).
Specifications
Specifications
| Antigen | MXI1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Mxi1 |
| Gene Accession No. | O09015, P50539, P50540 |
| Gene Alias | bHLHc11; class C basic helix-loop-helix protein 11; ENSMUSG00000067085; Gm10197; Mad2; MAX dimerization protein 2; Max interacting protein 1; MAX interactor 1; MAX interactor 1, dimerization protein; max-interacting protein 1; Max-interacting protein 1-like protein; Max-related transcription factor; MGC43220; MXD2; MXI; MXI1; MXI-WR; RP11-549L6.1 |
| Gene Symbols | Mxi1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.