Learn More
Invitrogen™ MXRA5 Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LMLSKDPRVS YQYRQDADEE ALYYTGVRAQ ILAEPEWVMQ PSIDIQLNRR QSTAKKVLLS YYTQYSQTIS TKDTRQARGR SWVMIEPSGA VQRDQTVLEG Highest antigen sequence indentity to the following orthologs: Mouse - 28%, Rat - 28%.
Specifications
Specifications
| Antigen | MXRA5 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MXRA5 |
| Gene Accession No. | Q9NR99 |
| Gene Alias | Adhesion protein with leucine-rich repeats and immunoglobulin domains related to perlecan; adlican; matrix remodeling associated 5; matrix-remodeling-associated protein 5; matrix-remodelling associated 5; MXRA5 |
| Gene Symbols | MXRA5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.