Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MXRA5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | MXRA5 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5121754
|
Invitrogen™
PA5121754 |
20 μL |
Each for $237.00
|
|
|||||
PIPA5121753
|
Invitrogen™
PA5121753 |
100 μL |
Each for $539.50
|
|
|||||
Description
Matrix-remodeling-associated protein 5 (MXRA5), also known as adhesion protein with leucine-rich repeats and immunoglobulin domains related to perlecan (Adlican), is a 2,828 amino acid secretory protein. Containing 12 Ig-like C2-type domains and 7 LRR repeats, MXRA5 is a member of the leucine-rich repeat and Ig domain-containing (LRRIG) family of proteins. MXRA5 expression, while decreasing with age from youth until late adulthood, increases from late adulthood through late elderhood. Most notably, NXRA5 has been shown to be overexpressed in centenarians. This expression pattern suggests a role of MXRA5 in the biology of aging and longevity.Specifications
MXRA5 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9NR99 | |
25878 | |
Pro2542-Phe2828 (Accession Q9NR99), with N-terminal His Tag. | |
Primary | |
-20°C | |
MXRA5 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Adhesion protein with leucine-rich repeats and immunoglobulin domains related to perlecan; adlican; matrix remodeling associated 5; matrix-remodeling-associated protein 5; matrix-remodelling associated 5; MXRA5 | |
MXRA5 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title