Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MXRA7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306326100UL
Description
MXRA7 Polyclonal antibody specifically detects MXRA7 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
MXRA7 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:500-1:3000 | |
FLJ41492, FLJ46603, HBV PreS1-transactivated protein 1, matrix-remodeling-associated protein 7, matrix-remodelling associated 7, PS1TP1, TMAP1, transmembrane anchor protein 1 | |
Produced in rabbits immunized with E. coli-derived Human MXRA7 fragment. | |
100 μg | |
Primary | |
Human | |
Purified |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH7.0 | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
439921 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction