Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MXRA8 Armenian Hamster anti-Human, Mouse, Clone: 9G2.D6, MilliporeSigma™

Catalog No. MABF2275100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2275100 100 μL
MABF227525 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2275100 Supplier MilliporeSigma Supplier No. MABF2275100UL
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

MXRA8 Monoclonal antibody specifically detects MXRA8 in Human, Mouse samples. It is validated for Flow Cytometry, Inhibition, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MXRA8
Applications Flow Cytometry, Inhibition Assays, Western Blot
Classification Monoclonal
Clone 9G2.D6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified Armenian Hamster monoclonal antibody IgG1 in PBS without azide.
Host Species Armenian Hamster
Immunogen Fc-fusion recombinant fragment corresponding to aa 22-336 of extracellular domain of mouse MXRA8 protein.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.