Learn More
Restek™ MXT Low Dead Volume Connector Kits for Metal Columns
SDP
Catalog No. 06611155 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
06-611-155 Stainless Steel
1 options
Catalog No. 06-611-155 Supplier Restek™ Supplier No. 20536
Connectors are Siltek treated to make them inert to active compounds, just like MXT columns. They can be used at temperatures up to 430°C without degrading the deactivated layer.
  • Connect a guard column/transfer line to an MXT stainless steel column
  • Low thermal mass tracks rapid oven temperature programming
  • Stainless steel ferrules and nuts
  • Siltek treated to ensure ultimate inertness
  • Available in Y and union configurations
  • Each kit includes: MXT Union, 1/32 in. Ferrules (2) and Nuts (2)
Specifications
Type Fitting/Connector
Includes MXT Union, 1/32 in. Ferrules (2) and Nuts (2)
Finish Siltek treated
For Use With (Application) 0.18/0.25/0.32 mm ID MXT Columns
Material Stainless Steel
Max. Operating Temperature (Metric) 430°C