Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ MXT Low Dead Volume Y Connector Kits for Metal Columns
SDP

Catalog No. 06611156 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Metal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
06-611-156 Metal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-611-156 Supplier Restek™ Supplier No. 20537
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Connectors are Siltek treated to make them inert to active compounds, just like MXT columns.Connect two MXT columns to one inlet or one MXT column to two detectors.

  • Siltek treated to make them inert to active compounds
  • Use with temperatures up to 430°C without degrading the deactivated layer
  • For 0.18/0.25/0.32 mm ID MXT Columns

Specifications

Type Y Fitting/Connector
Color Silver
For Use With (Application) 0.18/0.25/0.32 mm ID MXT Columns
Material Metal
Max. Operating Temperature (Metric) 430°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.