Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYADM Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS19143R

Description
MYADM, belonging to the MAL family, is a 322-amino acid protein having 7 hydrophobic transmembrane domains, several potential consensus sites for phosphorylation and sequence homology with the T-cell differentiation protein MAL and plasmolipin. It shares 75.8% sequence identity with mouse in the transmembrane regions and is associated with differentiation of hematopoietic cells or acute promyelocytic leukemia cells. Its expression is up-regulated in NB4 cells induced by all-trans retinoic acid whereas down regulation inhibits colony formation of bone marrow cells. MYADM could serve as a new marker for hematopoietic differentiation and is a target for diagnostic, prognostic and/or therapeutic interventions. It is known to express widely in brain and pancreatic islet cells.Specifications
MYADM | |
Polyclonal | |
Unconjugated | |
MYADM | |
MYADM; myeloid associated differentiation marker; myeloid upregulated protein; Myeloid up-regulated protein; Myeloid-associated differentiation marker; Protein SB135; SB135; UNQ553/PRO1110 | |
Rabbit | |
Protein A | |
RUO | |
91663 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH | |
Q96S97 | |
MYADM | |
KLH conjugated synthetic peptide derived from human MYADM . | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction