Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYB Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298885MFV610
Description
MYB Polyclonal antibody specifically detects MYB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MYB | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Cmyb;c-myb;c-myb_CDS;efg;Transcriptional activator Myb | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MYB | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
4602 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction