Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myb Monoclonal Antibody (1279CT309.289)
GREENER_CHOICE

Catalog No. PIMA537645
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537645 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537645 Supplier Invitrogen™ Supplier No. MA537645
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myb Monoclonal Antibody for Western Blot

Transcriptional activator; DNA-binding protein that specifically recognize the sequence 5'-YAAC[GT]G-3'. Plays an important role in the control of proliferation and differentiation of hematopoietic progenitor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myb
Applications Western Blot
Classification Monoclonal
Clone 1279CT309.289
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MYB
Gene Accession No. P10242
Gene Alias AI550390; Cmyb; c-myb; cmyb protein; c-myb_CDS; c-myb10A_CDS; c-myb13A_CDS; c-myb14A_CDS; c-myb8B_CDS; efg; gag-myb protein; M16449; MYB; myb proto-oncogene protein; MYB proto-oncogene, transcription factor; Mybl2; myb-related protein B; myeloblastosis oncogene; myeloblastosis oncogene-like 2; myeloblastosis proto-oncogene product; oncogene AMV; proto-oncogene c-Myb; RP1-32B1.1; transcription factor cmyb; transcriptional activator Myb; tumor-specific myb protein; v-myb avian myeloblastosis viral oncogene homolog
Gene Symbols MYB
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 1-185 amino acids from the Central region of human MYB.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4602
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.