Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYBBP1A Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10061050F
Description
MYBBP1A Polyclonal antibody specifically detects MYBBP1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MYBBP1A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
MYB binding protein (P160) 1a, myb-binding protein 1A, P160FLJ37886, p53-activated protein-2, PAP2 | |
The immunogen recognized by this antibody maps to a region between residue 450 and 500 of human MYB binding protein (P160) 1a using the numbering given in entry NP_055335.1 (GeneID 10514). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
10514 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction