Learn More
MYBBP1A Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00307

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Gorilla and Chimpanzee
The Myb family of transcription factors, which includes the structurally related A-, B-, and c-Myb genes, regulate differentiation, cellular growth and transactivating gene expression. c-Myb plays an essential role in controlling the proliferation and differentiation of hematopoietic cells, cellular levels of c-Myb decreasing as cells reach terminal differentiation. Myb-binding protein 1A (MYBBP1A) is a novel nuclear protein localized predomitly, but not exclusively, in nucleoli. Although initially isolated as a c-Myb-interacting protein, MYBBP1A is expressed ubiquitously, and associates with a number of different transcription factors. MYBBP1A associates with the aromatic hydrocarbon receptor (AhR), enhancing transactivation and increasing AhR-dependent gene expression. MYBBP1A is a powerful negative regulator of PPAR gamma coactivator 1 alpha (PGC-1 alpha) by p38 MAPK and also acts as a nucleocytoplasmic shuttling protein that utilizes CRM1-dependent and independent nuclear export pathways.Specifications
MYBBP1A | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9BQG0 | |
MYBBP1A | |
Between 450 and 500 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
MYBBP1A | |
Myb-binding protein 1A, P160 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10514 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.