Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYBPC1 Antibody (83B24), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250295G
Description
MYBPC1 Monoclonal specifically detects MYBPC1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MYBPC1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MYBPC1 | |
Crude preparation of human skeletal myosin | |
0.1 mL | |
Cellular Signaling | |
4604 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
83B24 | |
Western Blot | |
C-protein, skeletal muscle slow isoform, MYBPCC, MYBPCS, myosin binding protein C, slow type, myosin-binding protein C, slow-type, skeletal muscle C-protein, Slow MyBP-C, slow-type | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction