Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYBPC1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241119
Description
MYBPC1 Polyclonal specifically detects MYBPC1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MYBPC1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
MYBPC1 | |
Antibody was raised against a 12 amino acid synthetic peptide near the carboxy terminus of human MYBPC1. The immunogen is located within the last 50 amino acids of MYBPC1. Amino Acid Squence: LEGQQQSLHNKD | |
0.1 mg | |
Cellular Signaling | |
4604 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
ELISA, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml | |
C-protein, skeletal muscle slow isoform, MYBPCC, MYBPCS, myosin binding protein C, slow type, myosin-binding protein C, slow-type, skeletal muscle C-protein, Slow MyBP-C, slow-type | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction