Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYBPC1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $501.50
Specifications
Antigen | MYBPC1 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MYBPC1 Polyclonal specifically detects MYBPC1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
MYBPC1 | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
C-protein, skeletal muscle slow isoform, MYBPCC, MYBPCS, myosin binding protein C, slow type, myosin-binding protein C, slow-type, skeletal muscle C-protein, Slow MyBP-C, slow-type | |
MYBPC1 | |
IgG | |
Affinity Purified |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Cellular Signaling | |
PBS with 0.02% Sodium Azide | |
4604 | |
Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human MYBPC1. The immunogen is located within amino acids 160 - 210 of MYBPC1. Amino Acid Squence: DIRSAFKRSGEGQEDAGE | |
Primary | |
At least five isoforms of MYBPC1 are known to exist. MYBPC1 antibody is predicted not to cross-react with other MYBPC family members |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title