Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYBPC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101687 Supplier Invitrogen™ Supplier No. PA5101687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MYBPC1.

Myosin binding protein C (MYBPC) is a component of the thick filament of striated muscle, with the slow-type isoform designated MYBPC1. Both the fast-type (MYBPC2) and slow-type MYBPC protein contains seven immunoglobulin C2 motifs and three fibronectin type-III repeats. Multiple isoforms of MYBPC1 are known to exist, and are present in varying amounts in different skeletal muscles. It is thought that the MYBPC1 slow subfamily may play important roles in the assembly and stabilization of sarcomeric M- and A-bands and regulate the contractile properties of the actomyosin filaments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYBPC1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MYBPC1
Gene Accession No. Q00872, Q63518
Gene Alias 8030451F13Rik; C-protein, skeletal muscle slow isoform; LCCS4; muscle C-protein; MyBP-C slow; Mybpc1; MYBPCC; MYBPCS; myosin binding protein C, slow type; myosin binding protein C, slow-type; myosin-binding protein C, slow-type; skeletal muscle C-protein; slow MyBP-C; Slow-type C-protein
Gene Symbols MYBPC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MYBPC1(Accession Q00872), corresponding to amino acid residues A217-G267.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109272, 362867, 4604
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.