Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYBPC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583638
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583638 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583638 Supplier Invitrogen™ Supplier No. PA583638
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FKGKWLELGS KSGARFSFKE SHNSASNVYT VELHIGKVVL GDRGYYRLEV KAKDTCDSCG FNIDVEAPRQ DASGQSLESF KRTSEKKSDT A Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 73%.

The gene MYBPC2 belongs to the myosin-binding protein C family, which consists of fast-, slow-, and cardiac-type isoforms. These isoforms are myosin-associated proteins that are located in the C region of A bands in striated muscle. The protein encoded by MYBPC2 is the fast-type isoform. Mutations in genes encoding the slow-type and cardiac-type isoforms have been linked to distal arthrogryposis, type 1, and hypertrophic cardiomyopathy, respectively. MYBPC2-related diseases include Lethal Congenital Contracture Syndrome 4 and Nemaline Myopathy 9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYBPC2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYBPC2
Gene Accession No. Q14324
Gene Alias C-protein, skeletal muscle fast isoform; Fast MyBP-C; fast-type C-protein; fast-type muscle myosin-binding-protein C; MYBPC; Mybpc2; MYBPCF; myosin binding protein C, fast type; myosin binding protein C, fast-type; myosin-binding protein C, fast-type; mypc2; testicular tissue protein Li 126
Gene Symbols MYBPC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYBPC2. Recombinant protein control fragment (Product #RP-98873).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4606
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.