Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYBPC3 Monoclonal Antibody (OTI3E2), TrueMAB™, OriGene
SDP

Catalog No. 502543635
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3635 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3635 Supplier OriGene Supplier No. CF813746
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYBPC3 Monoclonal antibody specifically detects MYBPC3 in Human samples. It is validated for ELISA

Specifications

Antigen MYBPC3
Applications ELISA
Classification Monoclonal
Clone OTI3E2
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene MYBPC3
Gene Accession No. Q14896
Gene Alias cardiac C-protein; cardiac MyBP-C; cardiac myosin binding protein C; CMD1MM; CMH4; C-protein, cardiac muscle isoform; DKFZp779E1762; FHC; LVNC10; MYBP-C; Mybpc3; myosin binding protein C, cardiac; myosin binding protein C3; myosin-binding protein C, cardiac-type; truncated cardiac myosin-binding protein C
Host Species Mouse
Immunogen Full length human recombinant protein of human MYBPC3 C0-C1 (NP_000247) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4607
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.