Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYBPC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547925
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547925 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547925 Supplier Invitrogen™ Supplier No. PA547925
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse MyBPC3 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

Myosin binding protein C, cardiac-type (MYBPC3) is a thick filament-associated protein located in the crossbridge region of vertebrate striated muscle a bands. In vitro it binds MHC, F-actin and native thin filaments. It modifies the activity of actin-activated myosin ATPase. It may modulate muscle contraction or may play a more structural role. Mutations affecting the gene can result in cardiomyopathy - familial hypertrophic 4, cardiomyopathy - dilated 1MM, or left ventricular non-compaction 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYBPC3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene MYBPC3
Gene Accession No. Q14896
Gene Alias cardiac C-protein; cardiac MyBP-C; cardiac myosin binding protein C; CMD1MM; CMH4; C-protein, cardiac muscle isoform; DKFZp779E1762; FHC; LVNC10; MYBP-C; Mybpc3; myosin binding protein C, cardiac; myosin binding protein C3; myosin-binding protein C, cardiac-type; truncated cardiac myosin-binding protein C
Gene Symbols MYBPC3
Host Species Sheep
Immunogen E. coli-derived recombinant human MyBPC3 Arg1001-Lys1103.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4607
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.