Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYBPC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583689
Description
Immunogen sequence: TGDSDEWVFD KKLLCETEGR VRVETTKDRS IFTVEGAEKE DEGVYTVTVK NPVGEDQVNL TVKVIDVPDA.
Myosin binding protein C, cardiac-type (MYBPC3) is a thick filament-associated protein located in the crossbridge region of vertebrate striated muscle a bands. In vitro it binds MHC, F-actin and native thin filaments. It modifies the activity of actin-activated myosin ATPase. It may modulate muscle contraction or may play a more structural role. Mutations affecting the gene can result in cardiomyopathy - familial hypertrophic 4, cardiomyopathy - dilated 1MM, or left ventricular non-compaction 10.
Specifications
MYBPC3 | |
Polyclonal | |
Unconjugated | |
MYBPC3 | |
cardiac C-protein; cardiac MyBP-C; cardiac myosin binding protein C; CMD1MM; CMH4; C-protein, cardiac muscle isoform; DKFZp779E1762; FHC; LVNC10; MYBP-C; Mybpc3; myosin binding protein C, cardiac; myosin binding protein C3; myosin-binding protein C, cardiac-type; truncated cardiac myosin-binding protein C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4607 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14896 | |
MYBPC3 | |
Recombinant protein corresponding to Human MYBPC3. Recombinant protein control fragment (Product #RP-97333). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction