Learn More
Invitrogen™ Myc Tag Monoclonal Antibody
Mouse Monoclonal Antibody
$290.00 - $2345.00
Specifications
Antigen | Myc Tag |
---|---|
Concentration | ∼1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Description
NOTE: R950-25 was previously sold under product #46-0603. When re-ordering, please use #R950-25. This antibody recognizes amino acid sequence: Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu-. This product contains enough material for 25 Western blots. Western blot against 500 ng of recombinant protein gave a strong signal using chemiluminescent or alkaline phosphatase detection reagents. For Western blot, dilute in PBS containing 0.05% Tween-20 and 5% nonfat, dry milk (PBSTM).
Myc is a proto-oncogene and encodes a nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. The encoded protein forms a heterodimer with the related transcription factor MAX. This complex binds to the E box DNA consensus sequence and regulates the transcription of specific target genes. Amplification of this gene is frequently observed in numerous human cancers. Translocations involving this gene are associated with Burkitt lymphoma and multiple myeloma in human patients. There is evidence to show that translation initiates both from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site, resulting in the production of two isoforms with distinct N-termini.Specifications
Myc Tag | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG1 | |
Tag | |
MYC | |
46-0603; AU016757; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; MRTL; MYC; Myc protein; Myc proto-oncogene protein; Myc Tag; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; Niard; Nird; Proto-oncogene c-Myc; R95025; R950-25; transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog | |
MYC | |
Primary | |
Purified |
∼1 mg/mL | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 0.01% sodium azide | |
P01106 | |
4609 | |
c-Myc synthetic peptide: Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu-. | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.