Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myc Tag Monoclonal Antibody (A3-B4), Huabio

Catalog No. 502544769
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4769 Supplier HUABIO RESEARCH INC Supplier No. EM31105
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myc Tag Monoclonal antibody specifically detects Myc Tag in Tag samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen Myc Tag
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A3-B4
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias 46-0603; AU016757; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; MRTL; MYC; Myc protein; Myc proto-oncogene protein; Myc Tag; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; Niard; Nird; Proto-oncogene c-Myc; R95025; R950-25; transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog
Host Species Mouse
Immunogen Synthetic peptide immune sequence is EQKLISEEDLC-KLH.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.