Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYCBP Monoclonal Antibody (1D5D5)
GREENER_CHOICE

Catalog No. FEMA550265
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550265 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550265 Supplier Invitrogen™ Supplier No. MA550265
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYCBP Monoclonal Antibody for IHC (P), ELISA

The MYCBP gene encodes a protein that binds to the N-terminal region of MYC and stimulates the activation of E box-dependent transcription by MYC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYCBP
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1D5D5
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene MYCBP
Gene Accession No. Q99417
Gene Alias 5730488M09Rik; Amy1; AMY-1; Associate of Myc 1; associate of myc-1; AW552132; AW742590; c-myc binding protein; C-Myc-binding protein; hypothetical protein LOC539291; LOW QUALITY PROTEIN: C-Myc-binding protein; MYC binding protein; MYCBP
Gene Symbols MYCBP
Host Species Mouse
Immunogen Recombinant Human MYCBP Protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26292
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.