Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYCBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559476 Supplier Invitrogen™ Supplier No. PA559476
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVALYEEPEK PNSALDFLKH HLGAATPENP EIELLRLELA EMKEKYEAIV EE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

The MYCBP gene encodes a protein that binds to the N-terminal region of MYC and stimulates the activation of E box-dependent transcription by MYC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYCBP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYCBP
Gene Accession No. Q99417
Gene Alias 5730488M09Rik; Amy1; AMY-1; Associate of Myc 1; associate of myc-1; AW552132; AW742590; c-myc binding protein; C-Myc-binding protein; hypothetical protein LOC539291; LOW QUALITY PROTEIN: C-Myc-binding protein; MYC binding protein; MYCBP
Gene Symbols MYCBP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYCBP. Recombinant protein control fragment (Product #RP-99843).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26292
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.