Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ MYCBP Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596296
Description
Immunogen sequence: MAHYKAADSK REQFRRYLEK SGVLDTLTKV LVALYEEPEK PNSALDFLKH HLGAATPENP EIELLRLELA EMKEKYEAIV EENKKLKAKL AQYEPPQEEK RAE; Positive Samples: A-549, K-562; Cellular Location: Cytoplasm, Mitochondrion, Nucleus.
The MYCBP gene encodes a protein that binds to the N-terminal region of MYC and stimulates the activation of E box-dependent transcription by MYC.
            Specifications
| MYCBP | |
| Polyclonal | |
| Unconjugated | |
| MYCBP | |
| 5730488M09Rik; Amy1; AMY-1; Associate of Myc 1; associate of myc-1; AW552132; AW742590; c-myc binding protein; C-Myc-binding protein; hypothetical protein LOC539291; LOW QUALITY PROTEIN: C-Myc-binding protein; MYC binding protein; MYCBP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26292 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.95 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q99417 | |
| MYCBP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-103 of human MYCBP (NP_0364652). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction