Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYCBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599796 Supplier Invitrogen™ Supplier No. PA599796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MYCBP.

The MYCBP gene encodes a protein that binds to the N-terminal region of MYC and stimulates the activation of E box-dependent transcription by MYC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYCBP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MYCBP
Gene Accession No. Q99417, Q9EQS3
Gene Alias 5730488M09Rik; Amy1; AMY-1; Associate of Myc 1; associate of myc-1; AW552132; AW742590; c-myc binding protein; C-Myc-binding protein; hypothetical protein LOC539291; LOW QUALITY PROTEIN: C-Myc-binding protein; MYC binding protein; MYCBP
Gene Symbols MYCBP
Host Species Rabbit
Immunogen A synthesized peptide derived from human MYCBP(Accession Q99417), corresponding to amino acid residues M1-L31.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26292, 56309
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.