Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYCBP2 Mouse anti-Human, Clone: PM-8A9, MilliporeSigma™

Catalog No. MABN2397MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2397MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2397MI Supplier MilliporeSigma Supplier No. MABN2397
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MYCBP2 Antibody, clone PM-8A9 is a highly specific mouse monoclonal antibody that targets MYCBP2 (PAM) and has been tested in Immunoprecipitation and Western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYCBP2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone PM-8A9
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols MYCBP2,KIAA0916,PAM
Host Species Mouse
Immunogen MBP (Maltose-binding protein)-tagged recombinat fragment of 150 amino acids from the N-terminal region of human MYCBP2.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_055872
Test Specificity Clone PM-8A9 specifically detects MYCBP2 in HEK293 cells. It targets an epitope within the first 150 amino acids in the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.