Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYCBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584393 Supplier Invitrogen™ Supplier No. PA584393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CYHPAKPFQS QLPSVKEGIS EDLPVKMPCL YLQTLARHHH ENFVGYQDDN LFQDEMRYLR STSVPAPYIS VTPDASPNVF EEPESNMKSM PPSL Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 98%.

This gene encodes an E3 ubiquitin-protein ligase and member of the PHR (Phr1/MYCBP2, highwire and RPM-1) family of proteins. The encoded protein plays a role in axon guidance and synapse formation in the developing nervous system. In mammalian cells, this protein regulates the cAMP and mTOR signaling pathways, and may additionally regulate autophagy. Reduced expression of this gene has been observed in acute lymphoblastic leukemia patients and a mutation in this gene has been identified in patients with a rare inherited vision defect.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYCBP2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYCBP2
Gene Accession No. O75592
Gene Alias AU023734; AW107953; AW546647; C130061D10Rik; E3 ubiquitin-protein ligase MYCBP2; Highwire; KIAA0916; MYC binding protein 2; MYC binding protein 2, E3 ubiquitin protein ligase; Myc-binding protein 2; Mycbp2; Myc-bp2; PAM; pam, highwire, rpm 1; pam/highwire/rpm-1 protein; Phr; Phr1; probable E3 ubiquitin-protein ligase MYCBP2; protein associated with Myc; Protein Magellan; R75243
Gene Symbols MYCBP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYCBP2. Recombinant protein control fragment (Product #RP-108434).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23077
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.