Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYCBPAP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983024
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3024 Supplier Bioss Supplier No. BS13614R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MYCBPAP is a protein coding gene.

Specifications

Antigen MYCBPAP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MYCBPAP
Gene Accession No. Q8TBZ2
Gene Alias 4932408B01Rik; AMAM-1; Amap1; AMAP-1; AMY1-associating protein 1; AMY-1-binding protein 1; AW125474; cochlea derived protein of 106 kDa; Cod106; Mycbp associated protein; MYCBPAP; mycbpap {ECO:0000312; MYCBP-associated protein; Organ of Corti membrane-associated protein of 106 kDa; RGD:1359123}; testis secretory sperm-binding protein Li 214e
Gene Symbols MYCBPAP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MYCBPAP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84073
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.