Learn More
MYCL1/L-Myc Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304796AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Cellular DNA-binding proteins encoded by the c-myc genes. They are normally involved in nucleic acid metabolism and in mediating the cellular response to growth factors. Elevated and deregulated (constitutive) expression of c-myc proteins can cause tumoriSpecifications
MYCL1/L-Myc | |
Polyclonal | |
Unconjugated | |
MYCL | |
LMYC, MYCL, bHLHe38 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4610 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P12524 | |
MYCL | |
Between 314 and 364 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.